siRNA Details
| Gene Symbol: Samsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023380.2 | NP_075869.2 | 3 | 278 | ||
| GenBank mRNA | AF222928.1 | - | 3 | 234 | ||
| AK016072.1 | - | 1009 | ||||
| AK041340.1 | - | 3 | 210 | |||
| AK041566.1 | - | 3 | 253 | |||
| AK053946.1 | - | 3 | 253 | |||
| BC052906.1 | - | 3 | 290 | 30078141 | ||
| BC064778.1 | - | 3 | 278 | 30077237 | ||
| DQ333189.1 | - | 3 | 278 | |||

Samsn1
Genomic Map