siRNA Details
| Gene Symbol: Myl12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023402.2 | NP_075891.1 | 4 | 872 | ||
| GenBank mRNA | AK002885.1 | - | 4 | 874 | ||
| AK146115.1 | - | 4 | 890 | |||
| AK160123.1 | - | 4 | 915 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl12b| Gene Symbol: Myl12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023402.2 | NP_075891.1 | 4 | 872 | ||
| GenBank mRNA | AK002885.1 | - | 4 | 874 | ||
| AK146115.1 | - | 4 | 890 | |||
| AK160123.1 | - | 4 | 915 | |||