siRNA Details
| Gene Symbol: Ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141930.1 | NP_001135402.1 | 4 | 560 | ||
| NM_023465.4 | NP_075954.1 | 5 | 666 | |||
| GenBank mRNA | AB021261.1 | - | 5 | 583 | ||
| AK003595.1 | - | 4 | 562 | |||
| AK009066.1 | - | 4 | 487 | |||
| AK036181.1 | - | 5 | 668 | |||
| AK048258.1 | - | 5 | 664 | |||
| BC018436.1 | - | 5 | 626 | |||
| BC038253.1 | - | 5 | 642 | 4974951 | ||

Ctnnbip1
Genomic Map