siRNA Details
| Gene Symbol: TRPV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019841.6 | NP_062815.3 | 15 | 2447 | ||
| GenBank mRNA | AF304464.1 | - | 15 | 2532 | ||
| AJ487965.2 | - | 15 | 2185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRPV5| Gene Symbol: TRPV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019841.6 | NP_062815.3 | 15 | 2447 | ||
| GenBank mRNA | AF304464.1 | - | 15 | 2532 | ||
| AJ487965.2 | - | 15 | 2185 | |||