siRNA Details
| Gene Symbol: Cml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023493.2 | NP_075982.2 | 3 | 633 | ||
| GenBank mRNA | AF187099.1 | - | 1 | 118 | ||
| AK007530.1 | - | 3 | 633 | |||
| BC024605.1 | - | 3 | 616 | 4236653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cml5| Gene Symbol: Cml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023493.2 | NP_075982.2 | 3 | 633 | ||
| GenBank mRNA | AF187099.1 | - | 1 | 118 | ||
| AK007530.1 | - | 3 | 633 | |||
| BC024605.1 | - | 3 | 616 | 4236653 | ||