siRNA Details
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020389.2 | NP_065122.1 | 3 | 1021 | ||
GenBank mRNA | AJ272034.1 | - | 2 | 799 | ||
AJ421783.1 | - | 2 | 799 | |||
AJ549089.1 | - | 2 | 799 | |||
AY167926.1 | - | 2 | 799 | |||
BC128185.1 | - | 2 | 799 | 40113370 | ||
BC128186.1 | - | 2 | 799 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF139923, AK137418, AK135153, BC117995.
Panther Classification:
Gene Ontology Categories:
