siRNA Details
| Gene Symbol: Zfp120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023266.4 | NP_075755.4 | 3 | 1226 | ||
| NM_181266.2 | NP_851783.2 | 4 | 1290 | |||
| GenBank mRNA | AF211867.1 | - | 4 | 1290 | ||
| AF211868.1 | - | 4 | 1374 | |||
| AF211869.1 | - | 3 | 1226 | |||
| AF242378.1 | - | 3 | 1212 | |||
| AK033399.1 | - | 4 | 1287 | |||
| AK132318.1 | - | 4 | 1296 | |||
| BC013500.1 | - | 4 | 1293 | |||
| BC141128.1 | - | 4 | 1280 | 9055740 | ||

Zfp120
Genomic Map