siRNA Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290454.1 | NP_001277383.1 | 6 | 1502 | ||
| NM_023305.3 | NP_075794.3 | 7 | 1685 | |||
| GenBank mRNA | AF275549.3 | - | 7 | 1642 | ||
| AK009596.1 | - | 6 | 1461 | |||
| AK028477.1 | - | 7 | 1644 | |||
| AK028670.1 | - | 7 | 1636 | |||
| AK033121.1 | - | 7 | 1637 | |||
| AK051346.1 | - | 866 | ||||
| AK076078.1 | - | 7 | 1644 | |||
| AK212879.1 | - | 3 | 265 | |||
| BC065996.1 | - | 7 | 1633 | 5689710 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001012190, XM_008763626, BC083594.

Ubap1
Genomic Map