siRNA Details
| Gene Symbol: Agtpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048008.2 | NP_001041473.1 | 14, 15 | 2157 | ||
| NM_001284218.1 | NP_001271147.1 | 14, 15 | 2282 | |||
| NM_001284219.1 | NP_001271148.1 | 13, 14 | 2228 | |||
| NM_001284221.1 | NP_001271150.1 | 13, 14 | 2032 | |||
| NM_023328.3 | NP_075817.2 | 14, 15 | 2157 | |||
| GenBank mRNA | AF219141.1 | - | 14, 15 | 2096 | ||
| AK051467.1 | - | 13, 14 | 2068 | |||
| AK132695.1 | - | 14, 15 | 2097 | |||
| AK133215.1 | - | 14, 15 | 2216 | |||
| AK170046.1 | - | 14, 15 | 2100 | |||
| BC042475.1 | - | 13, 14 | 2228 | |||
| BC060633.1 | - | 14, 15 | 2201 | |||
| BC082335.1 | - | 13, 14 | 2020 | 30536788 | ||
| BM963763.1 | - | 1, 2 | 382 | |||

Agtpbp1
Genomic Map