siRNA Details
| Gene Symbol: Wfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023395.2 | NP_075884.1 | 7 | 1097 | ||
| GenBank mRNA | AK132541.1 | - | 7 | 1098 | ||
| AK142707.1 | - | 7 | 966 | |||
| AK161606.1 | - | 7 | 963 | |||
| BC025850.1 | - | 7 | 910 | |||
| BC038122.1 | - | 7 | 939 | |||
| BC132073.1 | - | 7 | 1026 | 40130350 | ||
| BC132469.1 | - | 7 | 1026 | 40130746 | ||
| BC145305.1 | - | 7 | 1023 | 9053859 | ||

Wfdc1
Genomic Map