siRNA Details
| Gene Symbol: Slc44a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023557.3 | NP_076046.1 | 4, 5 | 266 | ||
| GenBank mRNA | AK145621.1 | - | 4, 5 | 258 | ||
| BC010808.1 | - | 4, 5 | 264 | 4240658 | ||
| BC052652.1 | - | 4, 5 | 264 | 30034507 | ||

Slc44a4
Genomic Map