siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023617.2 | NP_076106.2 | 3 | 255 | ||
| GenBank mRNA | AF172276.2 | - | 3 | 331 | ||
| AK054065.1 | - | 3 | 214 | |||
| AK143048.1 | - | 3 | 235 | |||
| AK149434.1 | - | 3 | 216 | |||
| AY347569.1 | - | 3 | 132 | |||
| AY347570.1 | - | 3 | 132 | |||
| BC024046.1 | - | 3 | 205 | |||
| BC049853.1 | - | 3 | 207 | |||
| BC138876.1 | - | 3 | 234 | 8861898 | ||

Aox3
Genomic Map