siRNA Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023645.3 | NP_076134.1 | 2 | 481 | ||
| GenBank mRNA | AJ404004.1 | - | 2 | 243 | ||
| AK007835.1 | - | 2 | 474 | |||
| AK035391.1 | - | 2 | 474 | |||
| AK035899.1 | - | 2 | 482 | |||
| AK053315.1 | - | 2 | 474 | |||
| AK077468.1 | - | 1 | 1584 | |||
| AK079701.1 | - | 2 | 474 | |||
| AK170300.1 | - | 2 | 510 | |||
| BC023141.1 | - | 2 | 461 | 4018698 | ||

Kdelc1
Genomic Map