siRNA Details
| Gene Symbol: Pex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023651.4 | NP_076140.2 | 2 | 295 | ||
| GenBank mRNA | AF329877.2 | - | 2 | 204 | ||
| AJ304506.1 | - | 2 | 228 | |||
| AK011355.1 | - | 2 | 265 | |||
| AK029099.1 | - | 2 | 265 | |||
| AK029336.1 | - | 2 | 268 | |||
| AK031710.1 | - | 2 | 295 | |||
| AK031975.1 | - | 2 | 251 | |||
| AK153438.1 | - | 2 | 244 | |||
| AK169148.1 | - | 2 | 320 | |||
| AK182904.1 | - | 1 | 41 | |||
| BC023683.1 | - | 2 | 307 | 5343438 | ||

Pex13
Genomic Map