siRNA Details
| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023680.3 | NP_076169.2 | 7 | 659 | ||
| GenBank mRNA | AK012838.1 | - | 7 | 660 | ||
| AY046551.1 | - | 6 | 613 | |||
| AY165626.1 | - | 7 | 682 | |||
| AY165627.1 | - | 6 | 637 | |||
| BC076592.1 | - | 6 | 628 | |||

Tnfrsf22
Genomic Map