siRNA Details
| Gene Symbol: FXYD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164831.2 | NP_001158303.1 | 3 | 276 | ||
| NM_001164832.2 | NP_001158304.1 | 4 | 625 | |||
| NM_001164836.2 | NP_001158308.1 | 5 | 701 | |||
| NM_001164837.2 | NP_001158309.1 | 4 | 604 | |||
| NM_022003.3 | NP_071286.1 | 3 | 528 | |||
| GenBank mRNA | AK289629.1 | - | 3 | 468 | ||
| AK290757.1 | - | 5 | 403 | |||
| AK311736.1 | - | 3 | 140 | |||
| AL136699.1 | - | 3 | 114 | |||
| AL832811.1 | - | 4 | 221 | |||
| AY358976.1 | - | 3 | 138 | |||
| BC018652.2 | - | 5 | 311 | 2822864 | ||
| BC093040.1 | - | 3 | 138 | 6526843 | ||
| BT006717.1 | - | 2 | 83 | |||
| BX460202.2 | - | 4 | 273 | |||
| CR533457.1 | - | 2 | 83 | |||
| DA001721.1 | - | 3 | 116 | |||
| DA086584.1 | - | 3 | 220 | |||
| DA131017.1 | - | 3 | 232 | |||
| DA209740.1 | - | 3 | 529 | |||
| Gene Symbol: FXYD6-FXYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204268.1 | NP_001191197.1 | 3 | 528 | ||
| NM_001243598.2 | NP_001230527.1 | 3 | 528 | |||
| GenBank mRNA | CN310658.1 | - | 3 | 163 | ||
| FY211982.1 | - | 3 | 125 | |||
| FY214031.1 | - | 3 | 125 | |||

FXYD6
Genomic Map