siRNA Details
| Gene Symbol: Prl8a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023741.2 | NP_076230.1 | 5, 6 | 601 | ||
| GenBank mRNA | AF230921.1 | - | 5, 6 | 587 | ||
| AF230922.1 | - | 5, 6 | 590 | |||
| AF230923.1 | - | 6, 7 | 803 | |||
| AF466150.1 | - | 5, 6 | 602 | |||
| AK005568.1 | - | 5, 6 | 607 | |||
| AK167369.1 | - | 5, 6 | 608 | |||
| BC100326.1 | - | 5, 6 | 595 | 30787396 | ||

Prl8a8
Genomic Map