siRNA Details
| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023746.4 | NP_076235.1 | 2 | 196 | ||
| GenBank mRNA | AF226611.1 | - | 2 | 143 | ||
| AK005447.1 | - | 2 | 197 | |||
| BC051672.1 | - | 2 | 183 | 30017843 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl5a1| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023746.4 | NP_076235.1 | 2 | 196 | ||
| GenBank mRNA | AF226611.1 | - | 2 | 143 | ||
| AK005447.1 | - | 2 | 197 | |||
| BC051672.1 | - | 2 | 183 | 30017843 | ||