siRNA Details
| Gene Symbol: Yipf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023784.5 | NP_076273.1 | 4 | 472 | ||
| GenBank mRNA | AF217188.1 | - | 3 | 384 | ||
| AK009405.1 | - | 4 | 423 | |||
| BC089576.1 | - | 4 | 457 | 6758950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf7| Gene Symbol: Yipf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023784.5 | NP_076273.1 | 4 | 472 | ||
| GenBank mRNA | AF217188.1 | - | 3 | 384 | ||
| AK009405.1 | - | 4 | 423 | |||
| BC089576.1 | - | 4 | 457 | 6758950 | ||