siRNA Details
| Gene Symbol: Esm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023612.3 | NP_076101.1 | 1 | 459 | ||
| GenBank mRNA | AJ249354.1 | - | 1 | 322 | ||
| AK002913.1 | - | 1 | 353 | |||
| BC020038.1 | - | 1 | 332 | 4018658 | ||
| BC051919.1 | - | 1 | 459 | 6403364 | ||
| BC147648.1 | - | 1 | 327 | 9056638 | ||
| BC147650.1 | - | 1 | 327 | 9056640 | ||

Esm1
Genomic Map