siRNA Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023622.4 | NP_076111.2 | 4 | 460 | ||
| GenBank mRNA | AF237669.1 | - | 4 | 419 | ||
| AF239661.1 | - | 4 | 461 | |||
| AK004876.1 | - | 3 | 193 | |||
| AK053550.1 | - | 4 | 447 | |||
| AK134654.1 | - | 442 | ||||
| AK165023.1 | - | 4 | 448 | |||
| AK165746.1 | - | 4 | 453 | |||

Rgl3
Genomic Map