siRNA Details
| Gene Symbol: Ush1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163733.1 | NP_001157205.1 | 3, 4 | 282 | ||
| NM_001291182.1 | NP_001278111.1 | 3, 4 | 282 | |||
| NM_023649.2 | NP_076138.2 | 3, 4 | 282 | |||
| NM_153677.2 | NP_710143.2 | 3, 4 | 282 | |||
| GenBank mRNA | AF228924.1 | - | 3, 4 | 282 | ||
| AF228925.1 | - | 3, 4 | 282 | |||
| AY103465.1 | - | 3, 4 | 282 | |||
| AY246556.1 | - | 3, 4 | 256 | |||
| BC010819.1 | - | 3, 4 | 284 | 4217086 | ||

Ush1c
Genomic Map