siRNA Details
| Gene Symbol: Clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023671.2 | NP_076160.1 | 4 | 462 | ||
| GenBank mRNA | AK011789.1 | - | 4 | 450 | ||
| AK031877.1 | - | 4 | 444 | |||
| AK078050.1 | - | 4 | 418 | |||
| AK133981.1 | - | 4 | 455 | |||
| AK151356.1 | - | 4 | 448 | |||
| AK151944.1 | - | 4 | 448 | |||
| AK169259.1 | - | 4 | 426 | |||
| BC005575.1 | - | 4 | 428 | 3967933 | ||
| CX236388.1 | - | 4 | 462 | |||
| U53455.1 | - | 4 | 374 | |||

Clns1a
Genomic Map