siRNA Details
| Gene Symbol: Txnip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009935.2 | NP_001009935.1 | 1, 2 | 540 | ||
| NM_023719.2 | NP_076208.2 | 1, 2 | 537 | |||
| GenBank mRNA | AF173681.1 | - | 1, 2 | 304 | ||
| AF282826.1 | - | 1, 2 | 258 | |||
| AK004653.2 | - | 1, 2 | 537 | |||
| AK082476.1 | - | 1, 2 | 535 | |||
| AK088870.1 | - | 1, 2 | 535 | |||
| AK089304.1 | - | 1, 2 | 540 | |||
| AK089351.1 | - | 1, 2 | 538 | |||
| AK089403.1 | - | 1, 2 | 540 | |||
| AK155517.1 | - | 1, 2 | 538 | |||
| AK171205.1 | - | 1, 2 | 538 | |||
| AK171312.1 | - | 1, 2 | 538 | |||
| AK171425.1 | - | 1, 2 | 538 | |||
| AK171456.1 | - | 1, 2 | 535 | |||
| AK171464.1 | - | 1, 2 | 538 | |||
| AK171527.1 | - | 1, 2 | 538 | |||
| AK192038.1 | - | 1, 2 | 135 | |||
| AK214578.1 | - | 1, 2 | 112 | |||
| BC011212.1 | - | 1, 2 | 516 | 4193517 | ||
| BC031850.1 | - | 1, 2 | 515 | 3591421 | ||

Txnip
Genomic Map