siRNA Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023729.3 | NP_076218.3 | 3 | 337 | ||
| GenBank mRNA | AF459789.1 | - | 3 | 331 | ||
| AK016595.1 | - | 3 | 332 | |||
| BC058859.1 | - | 3 | 346 | 30011651 | ||
| BG088136.2 | - | 3 | 337 | |||

Asz1
Genomic Map