siRNA Details
| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023750.2 | NP_076239.1 | 5 | 2504 | ||
| GenBank mRNA | AK034259.1 | - | 5 | 2400 | ||
| AK083748.1 | - | 5 | 2445 | |||
| AK145426.1 | - | 6 | 2350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp84| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023750.2 | NP_076239.1 | 5 | 2504 | ||
| GenBank mRNA | AK034259.1 | - | 5 | 2400 | ||
| AK083748.1 | - | 5 | 2445 | |||
| AK145426.1 | - | 6 | 2350 | |||