siRNA Details
| Gene Symbol: Wdr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023790.2 | NP_076279.1 | 4, 5 | 551 | ||
| GenBank mRNA | AF053628.1 | - | 4, 5 | 539 | ||
| AK006536.1 | - | 4, 5 | 451 | |||
| AK154345.1 | - | 4, 5 | 542 | |||
| AK202399.1 | - | 3, 4 | 166 | |||
| BC083332.1 | - | 5, 6 | 456 | 6819838 | ||

Wdr54
Genomic Map