siRNA Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317395.1 | NP_001304324.1 | 7 | 1392 | ||
| NM_001317399.1 | NP_001304328.1 | 7 | 1320 | |||
| NM_031954.4 | NP_114160.1 | 7 | 1389 | |||
| NR_133897.1 | 6 | 1219 | ||||
| NR_133898.1 | 6 | 1175 | ||||
| GenBank mRNA | AB209358.1 | - | 2 | 3934 | ||
| AF113208.1 | - | 7 | 1134 | |||
| AK027543.1 | - | 7 | 1195 | |||
| AK056912.1 | - | 7 | 2797 | |||
| AK092170.1 | - | 6 | 1008 | |||
| AK123733.1 | - | 4 | 1626 | |||
| AK309783.1 | - | 6 | 1004 | |||
| AY205299.1 | - | 7 | 1195 | |||
| BC040062.1 | - | 7 | 1142 | 5494016 | ||

KCTD10
Genomic Map