siRNA Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290521.1 | NP_001277450.1 | 10 | 1231 | ||
| NM_001290522.1 | NP_001277451.1 | 10 | 1121 | |||
| NM_023597.2 | NP_076086.1 | 10 | 1124 | |||
| GenBank mRNA | AF274321.1 | - | 10 | 1101 | ||
| AK017259.1 | - | 10 | 1114 | |||
| AK141026.1 | - | 10 | 1229 | |||
| BC026500.1 | - | 9 | 1010 | |||
| BC037102.1 | - | 10 | 1114 | 5364307 | ||

Wdr31
Genomic Map