siRNA Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301230.1 | NP_001288159.1 | 7 | 1445 | ||
| NM_001301232.1 | NP_001288161.1 | 6 | 1338 | |||
| NM_023635.6 | NP_076124.1 | 6 | 1296 | |||
| GenBank mRNA | AF304376.1 | - | 6 | 1251 | ||
| AK010373.1 | - | 6 | 1214 | |||
| AK017093.1 | - | 1 | 1030 | |||
| AK030529.1 | - | 6 | 1243 | |||
| BC008173.1 | - | 7 | 1402 | 3710213 | ||
| BC009656.1 | - | 6 | 1249 | 3590089 | ||

Rab27a
Genomic Map