siRNA Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023645.3 | NP_076134.1 | 8, 9 | 1665 | ||
| GenBank mRNA | AJ404004.1 | - | 8, 9 | 1427 | ||
| AK007835.1 | - | 8, 9 | 1658 | |||
| AK035899.1 | - | 8, 9 | 1666 | |||
| AK053315.1 | - | 7, 8 | 1448 | |||
| AK079701.1 | - | 8, 9 | 1658 | |||
| AK170300.1 | - | 8, 9 | 1694 | |||

Kdelc1
Genomic Map