siRNA Details
| Gene Symbol: Dnaja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135112.1 | NP_001128584.1 | 4, 5 | 699 | ||
| NM_023646.4 | NP_076135.3 | 4, 5 | 699 | |||
| GenBank mRNA | AF325535.1 | - | 4, 5 | 740 | ||
| AK004575.1 | - | 4, 5 | 649 | |||
| AK004910.1 | - | 4, 5 | 651 | |||
| AK031250.1 | - | 4, 5 | 656 | |||
| AK167511.1 | - | 4, 5 | 661 | |||
| AY009320.1 | - | 4, 5 | 740 | |||
| BC003920.1 | - | 4, 5 | 632 | |||
| BC027240.1 | - | 4, 5 | 661 | 2654866 | ||

Dnaja3
Genomic Map