siRNA Details
| Gene Symbol: Nipa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256130.1 | NP_001243059.1 | 7 | 2272 | ||
| NM_001256131.1 | NP_001243060.1 | 6 | 2151 | |||
| NM_001256132.1 | NP_001243061.1 | 7 | 2076 | |||
| NM_001256133.1 | NP_001243062.1 | 5 | 1726 | |||
| NM_023647.6 | NP_076136.2 | 7 | 3026 | |||
| GenBank mRNA | AK028734.1 | - | 7 | 2984 | ||
| AK041427.1 | - | 7 | 2273 | |||
| AK154755.1 | - | 7 | 2230 | |||
| BC011415.2 | - | 1 | 1044 | |||

Nipa2
Genomic Map