siRNA Details
| Gene Symbol: Ndel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023668.2 | NP_076157.2 | 9 | 1749 | ||
| GenBank mRNA | AF290472.1 | - | 9 | 1681 | ||
| AF323918.1 | - | 9 | 1769 | |||
| AK146029.1 | - | 10 | 1713 | |||
| BC046796.1 | - | 9 | 1680 | 5718032 | ||

Ndel1
Genomic Map