siRNA Details
| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023695.2 | NP_076184.1 | 5 | 520 | ||
| GenBank mRNA | AF106853.1 | - | 5 | 520 | ||
| AK014012.1 | - | 5 | 522 | |||
| AK053869.1 | - | 5 | 521 | |||
| BC029012.1 | - | 4 | 504 | 5358714 | ||

Crybb1
Genomic Map