siRNA Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023831.3 | NP_076320.2 | 12 | 1976 | ||
| GenBank mRNA | AK032880.1 | - | 12 | 1977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift46| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023831.3 | NP_076320.2 | 12 | 1976 | ||
| GenBank mRNA | AK032880.1 | - | 12 | 1977 | ||