siRNA Details
| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023852.5 | NP_076341.1 | 5 | 721 | ||
| GenBank mRNA | AB232589.1 | - | 5 | 533 | ||
| AF312037.1 | - | 5 | 533 | |||
| AJ310532.1 | - | 5 | 533 | |||
| AK014132.1 | - | 5 | 643 | |||
| AK079574.1 | - | 5 | 618 | |||
| AK148904.1 | - | 5 | 642 | |||
| AK161006.1 | - | 5 | 619 | |||
| AK164316.1 | - | 5 | 666 | |||
| AY026947.1 | - | 5 | 602 | |||
| BC114335.2 | - | 5 | 594 | 40045744 | ||

Rab3c
Genomic Map