siRNA Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023876.3 | NP_076365.2 | 2 | 246 | ||
| GenBank mRNA | AJ276004.1 | - | 2 | 245 | ||
| AK020742.1 | - | 2 | 254 | |||
| AK034040.1 | - | 2 | 251 | |||
| AK034529.1 | - | 2 | 252 | |||
| AK037104.1 | - | 2 | 270 | |||
| AK039447.1 | - | 2 | 258 | |||
| AK039758.1 | - | 2 | 247 | |||
| AK079069.1 | - | 2 | 261 | |||
| AK168964.1 | - | 2 | 310 | |||

Elp4
Genomic Map