siRNA Details
| Gene Symbol: Tsc22d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023910.6 | NP_076399.4 | 2 | 960 | ||
| NM_029805.1 | NP_084081.1 | 2 | 960 | |||
| GenBank mRNA | AF201286.1 | - | 2 | 673 | ||
| AK018735.2 | - | 2 | 933 | |||
| AK040534.1 | - | 2 | 897 | |||
| AK079510.1 | - | 2 | 686 | |||
| AK143608.1 | - | 2 | 737 | |||
| AK144806.1 | - | 1 | 394 | |||
| AK150043.1 | - | 2 | 931 | |||
| AK154153.1 | - | 2 | 910 | |||
| AK154490.1 | - | 2 | 560 | |||
| AK155712.1 | - | 2 | 962 | |||
| AK170487.1 | - | 2 | 949 | |||
| BC018544.1 | - | 1 | 432 | 3666221 | ||
| BC023761.1 | - | 2 | 864 | 5343927 | ||
| BC145721.1 | - | 2 | 923 | 40131024 | ||
| BC145723.1 | - | 2 | 923 | 40131026 | ||

Tsc22d4
Genomic Map