siRNA Details
| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271386.1 | NP_001258315.1 | 5 | 360 | ||
| NM_024124.3 | NP_077038.2 | 3 | 419 | |||
| GenBank mRNA | AF235053.1 | - | 2 | 88 | ||
| AF279371.1 | - | 2 | 88 | |||
| AF324492.1 | - | 2 | 88 | |||
| AK087563.1 | - | 5 | 709 | |||
| AK133334.1 | - | 5 | 360 | |||
| BC098187.1 | - | 3 | 404 | 5363624 | ||
| EL604734.1 | - | 3 | 419 | |||

Hdac9
Genomic Map