siRNA Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024169.3 | NP_077131.2 | 6 | 697 | ||
| GenBank mRNA | AK003331.1 | - | 6 | 698 | ||
| AK007813.1 | - | 6 | 660 | |||
| BC002311.1 | - | 7 | 647 | |||
| BC022900.1 | - | 4 | 1266 | |||
| BC037596.1 | - | 6 | 653 | 5250344 | ||

Fkbp11
Genomic Map