siRNA Details
| Gene Symbol: Alg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024178.2 | NP_077140.1 | 2 | 236 | ||
| GenBank mRNA | AK011733.1 | - | 2 | 282 | ||
| BC002278.1 | - | 2 | 236 | 3496897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg14| Gene Symbol: Alg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024178.2 | NP_077140.1 | 2 | 236 | ||
| GenBank mRNA | AK011733.1 | - | 2 | 282 | ||
| BC002278.1 | - | 2 | 236 | 3496897 | ||