siRNA Details
| Gene Symbol: Mfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024200.4 | NP_077162.2 | 2 | 322 | ||
| GenBank mRNA | AK018181.1 | - | 2 | 319 | ||
| AK046313.1 | - | 2 | 275 | |||
| AK141611.1 | - | 2 | 323 | |||
| AK220169.1 | - | 2 | 310 | |||
| AY174062.1 | - | 1 | 451 | |||
| BC056641.1 | - | 2 | 281 | 5097345 | ||

Mfn1
Genomic Map