siRNA Details
| Gene Symbol: 2310033P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024210.2 | NP_077172.1 | 1 | 156 | ||
| GenBank mRNA | AK009601.1 | - | 1 | 143 | ||
| AK012628.1 | - | 1 | 167 | |||
| BC002181.1 | - | 1 | 156 | 3487283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310033P09Rik| Gene Symbol: 2310033P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024210.2 | NP_077172.1 | 1 | 156 | ||
| GenBank mRNA | AK009601.1 | - | 1 | 143 | ||
| AK012628.1 | - | 1 | 167 | |||
| BC002181.1 | - | 1 | 156 | 3487283 | ||