siRNA Details
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024213.2 | NP_077175.1 | 1, 2 | 266 | ||
GenBank mRNA | AK088759.1 | - | 1, 2 | 256 | ||
AK146000.1 | - | 1, 2 | 263 | |||
AK148625.1 | - | 1, 2 | 275 | |||
AK166061.1 | - | 1, 2 | 389 | |||
AK168031.1 | - | 1, 2 | 263 | |||
AK168897.1 | - | 1, 2 | 275 | |||
BC016237.1 | - | 1, 2 | 249 | 4504779 | ||
BC024870.1 | - | 1, 2 | 266 | 4949079 |
