siRNA Details
| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 1, 2 | 299 | ||
| GenBank mRNA | AK005247.1 | - | 1, 2 | 301 | ||
| AK014081.1 | - | 1, 2 | 249 | |||
| AK133585.1 | - | 1, 2 | 297 | |||
| AK156931.1 | - | 1, 2 | 234 | |||
| BC003465.1 | - | 1, 2 | 236 | 2648747 | ||

Zfp593
Genomic Map