siRNA Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024217.3 | NP_077179.1 | 4 | 644 | ||
| GenBank mRNA | AK003882.1 | - | 4 | 500 | ||
| AK079165.1 | - | 4 | 613 | |||
| AK172091.1 | - | 4 | 547 | |||
| AY241870.1 | - | 4 | 638 | |||
| BC003230.1 | - | 4 | 551 | 3483351 | ||
| BC145817.1 | - | 4 | 516 | 40131120 | ||
| BC145819.1 | - | 4 | 516 | 40131122 | ||

Cmtm3
Genomic Map