siRNA Details
| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024242.3 | NP_077204.2 | 2 | 657 | ||
| GenBank mRNA | AK017312.1 | - | 2 | 352 | ||
| AK152707.1 | - | 2 | 406 | |||
| AK161315.1 | - | 3 | 900 | |||
| BC006884.1 | - | 2 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Riok1| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024242.3 | NP_077204.2 | 2 | 657 | ||
| GenBank mRNA | AK017312.1 | - | 2 | 352 | ||
| AK152707.1 | - | 2 | 406 | |||
| AK161315.1 | - | 3 | 900 | |||
| BC006884.1 | - | 2 | 364 | |||