siRNA Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023876.3 | NP_076365.2 | 3 | 333 | ||
| GenBank mRNA | AJ276004.1 | - | 3 | 332 | ||
| AK020742.1 | - | 3 | 341 | |||
| AK034040.1 | - | 3 | 338 | |||
| AK034529.1 | - | 3 | 339 | |||
| AK037104.1 | - | 3 | 357 | |||
| AK039447.1 | - | 3 | 345 | |||
| AK039758.1 | - | 3 | 334 | |||
| AK079069.1 | - | 3 | 348 | |||
| AK168964.1 | - | 3 | 397 | |||

Elp4
Genomic Map