siRNA Details
| Gene Symbol: Asb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023906.3 | NP_076395.2 | 3 | 427 | ||
| GenBank mRNA | AF155354.1 | - | 2 | 242 | ||
| AK010315.1 | - | 3 | 363 | |||
| AK028489.1 | - | 4 | 533 | |||
| AK038086.1 | - | 3 | 408 | |||
| AK038897.1 | - | 4 | 491 | |||
| AK143580.1 | - | 3 | 362 | |||
| BB618690.1 | - | 3 | 429 | |||
| BC023086.1 | - | 3 | 361 | 4947610 | ||

Asb3
Genomic Map